SARS spike protein SARS Spike protein peptide (20 aa near C-terminus)
Artikelnr. PRSI3525P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:SARS Spike protein
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:20 aa near C-terminus
- Protein/peptide name:SARS Spike protein Peptide (20 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
SARS Spike peptide is used for blocking the activity of the SARS Spike antibody.