Order Entry
Austria
ContactUsLinkComponent
West nile virus WNV core protein peptide (15 aa near C-terminus)
West nile virus WNV core protein peptide (15 aa near C-terminus)
Catalog # PRSI3435P
Supplier:  ProSci Inc.
CAS Number:  
West nile virus WNV core protein peptide (15 aa near C-terminus)
Catalog # PRSI3435P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    West nile virus
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-Terminus
  • Protein/peptide name:
    WNV Core Protein Peptide (15 aa near C-Terminus)
  • Tested applications:
    Blocking

Specifications

About this item

West Nile virus peptide is used for blocking the activity of West Nile Virus Core Protein antibody.