West Nile virus (WNV) matrix protein peptide (15 aa near C-terminus)
Catalog # PRSI3431P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:West nile virus
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-Terminus
- Protein/peptide name:WNV Matrix Protein Peptide (15 aa near C-Terminus)
- Tested applications:Blocking
Specifications
About this item
West Nile virus peptide is used for blocking the activity of West Nile Virus Matrix Protein antibody.