- Antigen name:Tyrosine Hydroxylase
- Antigen symbol:TH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Pig,Monkey,Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TH-4|tyrosine hydroxylase|EC 1.14.16.2|TYH|Tyrosine 3-hydroxylase|Tyrosine 3-monooxygenase|TY3H|TH isoform 3|TH isoform a
- Immunogen:TH (Ser40) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phospho-Ser40 of rat tyrosine hydroxylase (TH).
- Size:100 µl
- Pk:100 µl
Tyrosine Hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines Dopamine and Norepinephrine. TH antibodies can therefore be used as markers for dopaminergic and noradrenergic neurons. We raised this polyclonal antibody against a peptide representing the sequence around Ser40 in rat TH. This antibody is suitable for most immunochemical applications in a variety of mammalian and some non-mammalian species.
Applications include Immunofluorescence (IF), Immunohistochemistry (IHC) and Western Blots (WB). Human, porcine, quail, mouse, and non-human primate have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for IF, IHC and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type:
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Monkey, Pig