Anti-KLRB1 Mouse Monoclonal Antibody
PRSI32-225
- Antibody type:Tag
- Antigen name:Killer cell lectin-like receptor subfamily B, member 1
- Antigen symbol:KLRB1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:NKRP1A|hNKR-P1A|NKR-P1A|NKR-P1|NKR|CLEC5B|CD161
- Immunogen:Recombinant human Thioredoxin (TRX).
- Size:100 µl
- Pk:100 µl
Thioredoxin (TRX) is a small ubiquitous protein (MW12 kDa) which is exist in a wide variety of prokaryotic and eukaryotic cells. Trx contains a redox active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site.This antibody is suitable for detecting fusion proteins which encode a Trx-Tag by immunoblotting and immunoprecipitation.The Monoclonal Antibody can detect a little Trx-Tag fusion proteins with negligible cross-reactivity with bacterial, insect, or mammalian lysates.
Ascitic fluid containing 0.03% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Tag
Antigen: KLRB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human