Anti-TOE1 Rabbit Polyclonal Antibody
PRSI30-814
- Antigen name:Target of EGR1, Member 1
- Antigen symbol:TOE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human TOE1.
- Size:50 µg
- Pk:50 µG
TOE1 belongs to the CAF1 family and 1 C3H1-type zinc finger. TOE1 inhibits cell growth rate and cell cycle. It induces CDKN1A expression as well as TGF-beta expression and mediates the inhibitory growth effect of EGR1.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TOE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat