Anti-DCP1A Rabbit Polyclonal Antibody
Product Details & Documents
Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway.Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DCP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Documents
10107-512.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:PRSI29-052
- Clonality:Polyclonal
- Reactivity:Human|Dog
- Form:Lyophilized
- Antigen synonyms:Smad4-interacting transcriptional co-activator|decapping enzyme |DCP1A|Nbla00360|HSA275986|putative protein product of Nbla00360|SMIF|decapping enzyme hDcp1a|transcription factor SMIF|SMAD4IP1
- Immunogen:A synthetic peptide corresponding a region of human DCP1A.
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:DCP1A
- Antigen name:Decapping mRNA 1A
- ELISA:Yes



