Anti-TLX2 Rabbit Polyclonal Antibody
PRSI29-030
- Antigen name:T-cell Leukemia Homeobox 2
- Antigen symbol:TLX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HOX11L1|T-cell leukemia homeobox protein 2|Homeobox protein Hox-11L1|NCX|TLX2
- Immunogen:A synthetic peptide corresponding a region of human TLX2.
- Size:100 µg
- Pk:100 µG
TLX2 contains 1 homeobox DNA-binding domain and the function remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TLX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat