Anti-TCP11L2 Rabbit Polyclonal Antibody
Catalog # PRSI27-043
Supplier: ProSci Inc.
Specifications
- Antigen name:T-Complex 11-like 2
- Antigen symbol:TCP11L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human TCP11L2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The specific function of TCP11L2 is not yet known.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TCP11L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human