Anti-GABARAPL2 Rabbit Polyclonal Antibody
Catalog # PRSI26-641
Supplier: ProSci Inc.
Specifications
- Antigen name:Gamma-aminobutyric acid receptor-associated protein-like 2
- Antigen symbol:GABARAPL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:GEF2|ATG8|ATG8C|GATE16|GEF-2|GATE-16
- Immunogen:A synthetic peptide corresponding a region of human GABARAPL2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GABARAPL2 belongs to the MAP1 LC3 family. It modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. The protein first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GABARAPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat