Anti-ANP32B Rabbit Polyclonal Antibody
PRSI26-635
- Antigen name:Acidic Nuclear Phosphoprotein 32 Family, Member B
- Antigen symbol:ANP32B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:APRIL|PHAPI2|SSP29
- Immunogen:A synthetic peptide corresponding a region of human ANP32B.
- Size:50 µg
- Pk:50 µG
ANP32B is a multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. ANP32B is required for the progression from the G1 to the S phase. ANP32B is an anti-apoptotic protein which functions as a caspase-3 inhibitor. It has no phosphatase 2A (PP2A) inhibitor activity.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human