Specifications
- Antigen symbol:HAO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human HAO2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
HAO2 is one of three related proteins that have 2-hydroxyacid oxidase activity yet differ in amino acid sequence, tissue expression and substrate preference. Subcellular location of the protein is the peroxisome. Specifically, the protein is expressed predominantly in liver and kidney and has the highest activity toward the substrate 2-hydroxypalmitate. Two alternatively spliced variants encoding the same isoform have been described.This gene is one of three related genes that have 2-hydroxyacid oxidase activity yet differ in encoded protein amino acid sequence, tissue expression and substrate preference. Subcellular location of the encoded protein is the peroxisome. Specifically, this gene is expressed predominantly in liver and kidney and has the highest activity toward the substrate 2-hydroxypalmitate. Two alternatively spliced variants encoding the same isoform have been described.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HAO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human