Anti-ACSL1 Rabbit Polyclonal Antibody
PRSI7081
- Antigen name:Acyl-CoA Synthetase Long-chain Family Member 1
- Antigen symbol:ACSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:FACL1|FACL2|LACS|LACS1|LACS2|ACS1
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:Rabbit polyclonal ACSL1 antibody was raised against an 18 amino acid peptide near the center of human ACSL1.
- Tested applications:E, WB, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
ACSL1 Antibody: Long-chain acyl coenzyme A synthetase 1 (ACSL1) catalyzes the synthesis of acyl-CoA from long-chain fatty acids in an ATP-dependent manner. ACSL1 is a member of a family of long-chain acyl-CoA synthetases which differ in substrate preference, tissue expression, and subcellular localization. In mouse, ASCL1 is the major acyl-CoA enzyme in the heart, providing 60-90% of heart ATP. Loss of ASCL1 either globally or in heart ventricles resulted in impaired fatty acid oxidation, activation of the mammalian target of rapamycin (mTOR), and cardiac hypertrophy.
Type:
Antigen: ACSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat