Anti-SLAIN2 Rabbit Polyclonal Antibody
PRSI5471
- Antigen name:SLAIN Motif Family Member 2
- Antigen symbol:SLAIN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Antigen synonyms:SLAIN motif-containing protein 2|KIAA1458
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SLAIN2 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human SLAIN2.
- Tested applications:E, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
SLAIN2 Antibody: SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Type:
Antigen: SLAIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human