Specifications
- Antigen name:Muscular LMNA-interacting protein
- Antigen symbol:MLIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:C6orf142|MLIP|Muscular-enriched A-type laminin-interacting protein|Muscular LMNA-interacting protein
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:MLIP antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human MLIP.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MLIP Antibody: The muscle-enriched A-type lamin-interacting protein (MLIP) is ubiquitously expressed, although most predominantly in heart, skeletal and smooth muscle, and interacts directly and co-localizes with lamin A and C in the nuclear envelope. MLIP also co-localizes with promyelocytic leukemia (PML) bodies within the nucleus. Decreasing lamin A/C expression with siRNA results in the upregulation and mislocalization of MLIP, suggesting that MLIP localization to the nuclear envelope is dependent on the lamin proteins. It has been suggested that MLIP may function as a tethering protein linking LMNA to factors within the nuclear envelope of within PML bodies
Type:
Antigen: MLIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human