- Antigen name:Leucine Rich Repeat And Fibronectin Type III Domain Containing 2
- Antigen symbol:LRFN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:Leucine-rich repeat and fibronectin type-III domain-containing protein 2|SALM1|Synaptic adhesion-like molecule 1.|fibronectin type III|immunoglobulin and leucine rich repeat domains 2 FIGLER2|LRFN2|LRFN2_HUMAN|leucine rich repeat and fibronectin type III domain containing 2|RP11-535K1.2
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:LRFN2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human LRFN2.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
LRFN2 Antibody: LRFN2 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
Type:
Antigen: LRFN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat