- Antigen name:Transmembrane channel-like protein 8
- Antigen symbol:TMC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:EV2|EVIN2|EVER2
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:EVER2 antibody was raised against a 14 amino acid synthetic peptide from near the center of human EVER2.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
EVER2 Antibody: Epidermodysplasia verruciformis (EV) is an autosomal recessive genodermatosis associated with a high risk of skin cancers resulting from a high susceptibility to infection by specific human papillomaviruses. Mutations in two homologous genes EVER1 and EVER2 cause the majority of EV cases. These two proteins form a complex and interact with the zinc transporter ZnT-1 in the endoplasmic reticulum. Cells lacking EVER2 accumulated higher levels of zinc in the nucleolus and nucleus compare to those cells with and intact EVER2 gene, indicating that one role of EVER2 is to regulate the intracellular distribution of zinc.
Type:
Antigen: TMC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat