Order Entry
Austria
ContactUsLinkComponent
Brimonidine Related Compounds, Reference Standard
Brimonidine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Brimonidine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    50358-63-9
  • Synonym(s):
    5-Bromoquinoxalin-6-amine
  • Formula:
    C₈H₆BrN₃
  • Description:
    Brimonidine Related Compound A

Specifications

Frequently Bought Together