Order Entry
Austria
Orders LinkContactUsLinkComponent
Vancomycin, Reference Standards

Vancomycin, Reference Standards

Supplier:   USP
Vancomycin, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1709007.
  • 1709018.
  • 1709029.
  • Description:
  • Vancomycin Hydrochloride
  • Vancomycin B with Monodechlorovancomycin
  • Vancomycin B
  • CAS No.:
  • 1404-93-9
  • 1404-93-9
  • 1404-93-9
  • UN No.:
  • 3288

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1709007.
    Vancomycin Hydrochloride
    Vancomycin Hydrochloride
    USP reference standard
    Vial
    4
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 388,00 €
    388,00 €
    1 * 500 mg
       388,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1709007.
    Description
    Vancomycin Hydrochloride
    CAS No.
    1404-93-9
    UN No.
  • 1709018.
    Vancomycin B with Monodechlorovancomycin
    Vancomycin B with Monodechlorovancomycin
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 1 300,00 €
    1 300,00 €
    1 * 300 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1709018.
    Description
    Vancomycin B with Monodechlorovancomycin
    CAS No.
    1404-93-9
    UN No.
  • 1709029.
    Vancomycin B
    Vancomycin B
    USP reference standard
    Vial
    350 mg
    Log in for availability
    Log in for availability
    1 * 350 mg
    1 * 350 mg - 369,00 €
    369,00 €
    1 * 350 mg
       369,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1709029.
    Description
    Vancomycin B
    CAS No.
    1404-93-9
    UN No.
    3288