Safety lockouts, dual level
: Brady
Especially designed to allow for the removal of personal padlocks these lockouts enable a supervisory padlock (or two padlocks on the 12 holes) to remain safely locked in place, ensuring that the machine remains isolated.
- Foolproof access to the hasp entry and a tamperproof locking facility adds another degree of safety
- Manufactured from marine grade 316 stainless steel
- 5 mm shackle will fit most electrical switch gear, cabinets and distribution boards
Shackle W×D×H: 35×5×35 mm
: Available in 6 hole×1 supervisory, or 12 holes×2 supervisory configurations.
Frequently Bought Together