ContactUsLinkComponent
Compact circuit breaker lockout
Compact circuit breaker lockout
Catalog # BRDY836597
Supplier:  Brady
undefined
Compact circuit breaker lockout
Catalog # BRDY836597
Supplier:  Brady
Specifications
  • Type:
    Circuit breaker lockout
  • Colour:
    Red
  • Pk:
    1
Specifications
About this item
Compact universal design to lock out a large range of single, double and triple-pole circuit breakers.
Place lockout over circuit breaker handle, tighten set screw and attach padlock and tag.