Accessories for ARE/AREX-6 digital and digital PRO hotplate stirrers
Frequently Bought Together
Probes for 108 (Testo)
: Testo
VWR®, Syringe Filters
: VWR Collection
VWR®, Storage Boxes
: VWR Collection
Acetic acid glacial USP, Ph. Eur.
: VWR Chemicals
VWR®, Tip Boxes, Empty
: VWR Collection
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Safety spectacles, Viper
: Bollé Safety
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® Advanced Magnetic Hotplate Stirrers
: VWR Collection
VWR®, Beakers, Low Form
: VWR Collection
VWR®, Cover Glasses, Square
: VWR Collection
Funnels, drum
: VITLAB
Ammonia solution 28%, GPR RECTAPUR®
: VWR Chemicals
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
: Honeywell Chemicals
Water standards for Karl Fischer titration, Hydranal™
: Honeywell Chemicals
Copper etchant
: Thermo Fisher Scientific