Spill containment pop-up pools
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
The space-saving and lightweight pools are designed to collect spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery. The durable PVC coated vinyl fabric is 20% heavier than other models, providing increased abrasion resistance and reliable seam adhesions.
- Folds tightly and stows easily
- Yellow colour provides visibility and easy identification
- Embedded foam ring top raises containment wall as fluid level increases
Certifications: Helps to comply with EPA for containment and spill prevention (SPCC).
Frequently Bought Together