Order Entry
Austria
ContactUsLinkComponent
Piroctone Related Compounds, Reference Standard
Piroctone Related Compounds, Reference Standard
Supplier:  USP
Certificates
Piroctone Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    68890-66-4
  • UN No.:
    3077
  • Formula:
    C₁₆H₃₀N₂O₃
  • Description:
    Piroctone Olamine
  • Synonym(s):
    4-methyl-6-(2,4,4-trimethylpentyl)-2H-pyran-2-one

Specifications

Frequently Bought Together