Anti-HTRA3 Rabbit Polyclonal Antibody
Catalog # BSBTA05478
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Serine protease HTRA3
- Antigen symbol:HTRA3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine protease HTRA3(HTRA3) detection. Tested with WB in Human.
Type: Primary
Antigen: HTRA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human