Phosphatase assay kit, BIOMOL® Green
BIOMOL® Green provides a simple and convenient method for colorimetric phosphate quantitation. It is suitable for applications include assays for phospholipid phosphatases, tyrosyl-tRNA synthetase (coupled with pyrophosphatase) and a viral RNA triphosphatase. It is most widely applied to protein phosphatase assays.
- Convenient one-step reagent, no mixing needed
- Extremely stable with a long shelf-life, >6 months at +4 °C
- Excellent sensitivity, ~50 pmol in 100 µl
- Can be used in cuvette or microplate-based assays, making it ideal for high-throughput applications
: Kit contains Biomol® Green reagent and Phosphate standard.
Frequently Bought Together