Order Entry
Austria
ContactUsLinkComponent
Anagrelide, Reference Standards
Anagrelide, Reference Standards
Supplier:  USP
Certificates
Anagrelide, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    823178-43-4
  • Formula:
    C₁₀H₇Cl₂N₃O,HCl, H₂O
  • Description:
    Anagrelide Hydrochloride
  • Synonym(s):
    Ethyl 2-(6-amino-2,3-dichlorobenzylamino)acetate

Specifications

Frequently Bought Together