Order Entry
Austria
Orders LinkContactUsLinkComponent
Temozolomide, Reference Standards

Temozolomide, Reference Standards

Supplier:   USP
Temozolomide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1643554.
  • 1643565.
  • 1643576.
  • 1643543.
  • Description:
  • Temozolomide Related Compound A
  • Temozolomide Related Compound B
  • Temozolomide Related Compound C
  • Temozolomide
  • Synonym(s):
  • 4-Diazo-4H-imidazole-5-carboxamide
  • 3-Methyl-1,1-diphenylurea
  • diphenylcarbamic chloride
  • CAS No.:
  • 7008-85-7
  • 13114-72-2
  • 83-01-2
  • 85622-93-1
  • Formula:
  • C₄H₃N₅O
  • C₁₄H₁₄N₂O
  • C₁₃H₁₀ClNO
  • C₆H₆N₆O₂
  • UN No.:
  • 3261

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1643554.
    Temozolomide Related Compound A
    Temozolomide Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 270,00 €
    1 270,00 €
    1 * 15 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643554.
    Description
    Temozolomide Related Compound A
    Synonym(s)
    4-Diazo-4H-imidazole-5-carboxamide
    CAS No.
    7008-85-7
    Formula
    C₄H₃N₅O
    UN No.
  • 1643565.
    Temozolomide Related Compound B
    Temozolomide Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 150,00 €
    1 150,00 €
    1 * 15 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643565.
    Description
    Temozolomide Related Compound B
    Synonym(s)
    3-Methyl-1,1-diphenylurea
    CAS No.
    13114-72-2
    Formula
    C₁₄H₁₄N₂O
    UN No.
  • 1643576.
    Temozolomide Related Compound C
    Temozolomide Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 620,00 €
    620,00 €
    1 * 15 mg
       620,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643576.
    Description
    Temozolomide Related Compound C
    Synonym(s)
    diphenylcarbamic chloride
    CAS No.
    83-01-2
    Formula
    C₁₃H₁₀ClNO
    UN No.
    3261
  • 1643543.
    Temozolomide
    Temozolomide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 891,00 €
    891,00 €
    1 * 200 mg
       891,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643543.
    Description
    Temozolomide
    Synonym(s)
    CAS No.
    85622-93-1
    Formula
    C₆H₆N₆O₂
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.