Order Entry
Austria
Orders LinkContactUsLinkComponent
Sevoflurane, Reference Standards

Sevoflurane, Reference Standards

Supplier:   USP
Sevoflurane, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1612550.
  • 1612572.
  • 1612594.
  • 1612540.
  • Description:
  • Sevoflurane Related Compound A
  • Sevoflurane Related Compound B
  • Sevoflurane Related Compound C
  • Sevoflurane
  • Synonym(s):
  • 1,1,1,3,3-Pentafluoroisopropenyl fluoromethyl ether
  • 1,1,1,3,3,3-hexafluoro-2-methoxy-propane
  • 1,1,1,3,3,3-hexafluoro-2-propanol
  • CAS No.:
  • 58109-34-5
  • 13171-18-1
  • 920-66-1
  • 28523-86-6
  • Formula:
  • C₄H₂F₆O
  • C₄H₄F₆O
  • C₃H₂F₆O
  • C₄H₃F₇O
  • UN No.:
  • 2810
  • 3334
  • 2922
  • 3334

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1612550.
    Sevoflurane Related Compound A
    Sevoflurane Related Compound A
    USP reference standard
    Ampule
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL - 1 300,00 €
    1 300,00 €
    1 * 0,2 mL
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1612550.
    Description
    Sevoflurane Related Compound A
    Synonym(s)
    1,1,1,3,3-Pentafluoroisopropenyl fluoromethyl ether
    CAS No.
    58109-34-5
    Formula
    C₄H₂F₆O
    UN No.
    2810
  • 1612572.
    Sevoflurane Related Compound B
    Sevoflurane Related Compound B
    USP reference standard
    Ampule
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL - 1 290,00 €
    1 290,00 €
    1 * 0,2 mL
       1 290,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1612572.
    Description
    Sevoflurane Related Compound B
    Synonym(s)
    1,1,1,3,3,3-hexafluoro-2-methoxy-propane
    CAS No.
    13171-18-1
    Formula
    C₄H₄F₆O
    UN No.
    3334
  • 1612594.
    Sevoflurane Related Compound C
    Sevoflurane Related Compound C
    USP reference standard
    Ampule
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL - 1 290,00 €
    1 290,00 €
    1 * 0,2 mL
       1 290,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1612594.
    Description
    Sevoflurane Related Compound C
    Synonym(s)
    1,1,1,3,3,3-hexafluoro-2-propanol
    CAS No.
    920-66-1
    Formula
    C₃H₂F₆O
    UN No.
    2922
  • 1612540.
    Sevoflurane
    Sevoflurane
    USP reference standard
    Ampule
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 412,00 €
    412,00 €
    1 * 1 mL
       412,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1612540.
    Description
    Sevoflurane
    Synonym(s)
    CAS No.
    28523-86-6
    Formula
    C₄H₃F₇O
    UN No.
    3334