Order Entry
Austria
ContactUsLinkComponent
Oxymetazoline Related Compounds, Reference Standard
Oxymetazoline Related Compounds, Reference Standard
Supplier:  USP
Certificates
Oxymetazoline Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    1391053-50-1
  • Synonym(s):
    N-(2-Aminoethyl)-2-[4-(tert-butyl)-3-hydroxy-2,6-dimethylphenyl]acetamide hydrochloride
  • Formula:
    C₁₆H₂₇ClN₂O₂
  • Description:
    Oxymetazoline Related Compound A
  • UN No.:
    2811

Specifications

Frequently Bought Together