ContactUsLinkComponent
Chloride FS
Chloride FS
Catalog # DIAS112009990314
CAS Number:  
Chloride FS
Catalog # DIAS112009990314
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Chloride FS, 6×25 ml
  • Environmentally Preferable:
Specifications
About this item
Diagnostic reagent for quantitative in vitro determination of chloride in serum or plasma by photometric test using thiocyanate.