Order Entry
Austria
ContactUsLinkComponent
Epinephrine, Reference Standards
Epinephrine, Reference Standards
Supplier:  USP
Certificates
Epinephrine, Reference Standards
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    51-43-4
  • UN No.:
    2811
  • Description:
    Epinephrine
  • Formula:
    C₁₂H₁₉NO₁₀

Specifications

Related Information

Frequently Bought Together