Acrylamide/Bis-Acrylamide
Premix of acrylamide with the common cross-linking reagent of final ratio 29:1.
- Soluble in water
- DNA sequencing gels and protein separation
Frequently Bought Together
Premix of acrylamide with the common cross-linking reagent of final ratio 29:1.