Specifications
- Packed:Sheet
- Colour:Blue
- L×W:540×410 mm
- Environmentally Preferable:
- Pk:50
Specifications
About this item
The sterile Chemoprotect® prep mat has been specially developed for cytotoxic drug preparation. The threelayer structure of the material allows safe working and risk-free subsequent disposal. The prep mat is available sterile or non-sterile in two sizes for most workplaces.
- Lint-free fluid-permeable surface
- Fluid-retaining, highly absorbent middle layer
- Fluid-impermeable bottom layer