Order Entry
Austria
Orders LinkContactUsLinkComponent
Acitretin, Reference Standards

Acitretin, Reference Standards

Supplier:   USP
Acitretin, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1011007.
  • 1011018.
  • 1011029.
  • Description:
  • Acitretin
  • Acitretin Related Compound A
  • Acitretin Related Compound B
  • Synonym(s):
  • (2Z,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
  • ethyl (all-E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoate
  • CAS No.:
  • 55079-83-9
  • 69427-46-9
  • 54350-48-0
  • Formula:
  • C₂₁H₂₆O₃
  • C₂₁H₂₆O₃
  • C₂₃H₃₀O₃
  • UN No.:
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pk
    Availability
    Price
  • 1011007.
    Acitretin
    Acitretin
    USP reference standard
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 807,00 €
    807,00 €
    1 * 200 mg
       807,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1011007.
    Description
    Acitretin
    Synonym(s)
    CAS No.
    55079-83-9
    Formula
    C₂₁H₂₆O₃
    UN No.
    3077
  • 1011018.
    Acitretin Related Compound A
    Acitretin Related Compound A
    USP reference standard
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 200,00 €
    1 200,00 €
    1 * 20 mg
       1 200,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1011018.
    Description
    Acitretin Related Compound A
    Synonym(s)
    (2Z,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
    CAS No.
    69427-46-9
    Formula
    C₂₁H₂₆O₃
    UN No.
  • 1011029.
    Acitretin Related Compound B
    Acitretin Related Compound B
    USP reference standard
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 190,00 €
    1 190,00 €
    1 * 20 mg
       1 190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1011029.
    Description
    Acitretin Related Compound B
    Synonym(s)
    ethyl (all-E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoate
    CAS No.
    54350-48-0
    Formula
    C₂₃H₃₀O₃
    UN No.
    3077
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.