Order Entry
Austria
ContactUsLinkComponent
 
Sodium standard solution, NIST-3152A
Catalog # PROONIST-3152A
CAS Number:  
Sodium standard solution, NIST-3152A
Catalog # PROONIST-3152A
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pk:
    50 mL
  • Description:
    NIST-3152A
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item

This standard reference material is intended for use as a primary calibration standard for the quantitative determination of sodium.

A unit consists of 50 ml of an aqueous solution in a high-density polyethylene bottle sealed in an aluminised bag.

Certifications: Traceable to NIST and SRM numbers.