Order Entry
Austria
ContactUsLinkComponent
Iron standard solution
Iron standard solution
Catalog # PROONIST-3126A
CAS Number:  
Iron standard solution
Catalog # PROONIST-3126A
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃
  • Pk:
    50 mL
  • Description:
    NIST-3126A Iron standard solution
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

This standard reference material is intended for use as a primary calibration standard for the quantitative determination of iron.

A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.

Certifications: Traceable to NIST and SRM numbers.