Order Entry
Austria
ContactUsLinkComponent
 
Detergent for manual cleaning of glassware, powder, neodisher® PM 5
  141-0941
undefined
Detergent for manual cleaning of glassware, powder, neodisher® PM 5
  141-0941

 

  • Description:
    neodisher® PM 5
  • Seq:
    05
  • Packed:
    3 kg bucket
  • Pk:
    4

 

 

For manual cleaning of laboratory glassware and surgical instruments in hospitals, medical and biological laboratories, and food industry laboratories.

  • Mildly alkaline powder for washing surgical instruments and laboratory glassware by hand
  • Components: <5% non-ionic surfactants, anionic surfactants, bleaching agents (percarbonates); >30% phosphates
  • Dosage: 0,5 to 2%