Order Entry
Austria
ContactUsLinkComponent
Nickel standard solutions
Nickel standard solutions
Artikelnr. PROONIST-3136
CAS-Nummer:  
Nickel standard solutions
Artikelnr. PROONIST-3136
CAS-Nummer:  

Spezifikationen

  • Concentration:
    1%
  • Matrix:
    10% HNO₃
  • Pack type:
    Ampoule
  • Pk:
    5
  • Description:
    NIST-3136
  • Element:
    Nickel
  • Symbol:
    Ni

Spezifikationen

Über diesen Artikel

This reference standard material is intended for use as a primary calibration standard for the quantitative determination of nickel.

A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of nickel.

Certifications: Traceable to NIST and SRM numbers.