Order Entry
Austria
ContactUsLinkComponent
Copper standard solution, NIST-3114
Copper standard solution, NIST-3114
Catalog # PROONIST-3114
CAS Number:  
Copper standard solution, NIST-3114
Catalog # PROONIST-3114
CAS Number:  

Specifications

  • Concentration:
    10 mg/g
  • Matrix:
    10% HNO₃
  • Pk:
    5
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item

This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of copper.

A unit consists of 50 ml of an aqueous solution in a high-density polyethylene bottle sealed in an aluminised bag.

Certifications: Traceable to NIST and SRM numbers.