Order Entry
Austria
ContactUsLinkComponent
Cadmium standard solution, NIST-3108
Cadmium standard solution, NIST-3108
Catalog # PROONIST-3108
CAS Number:  
Cadmium standard solution, NIST-3108
Catalog # PROONIST-3108
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    10% HNO₃
  • Pk:
    50 mL
  • Description:
    NIST-3108
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

This reference standard is intended for use as a primary calibration standard for the quantitative determination of cadmium.

A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.

Certifications: Traceable to NIST and SRM numbers.