Beryllium standard solution
Artikelnr. PROONIST-3105A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:10% HNO₃
- Pk:5
- Description:NIST-3105A
- Element:Beryllium
- Symbol:Be
Specifications
About this item
This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of beryllium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of beryllium.
Certifications: Traceable to NIST and SRM numbers.