Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, sulphadiazines, Ph. Eur., RS
Organic reference standards, sulphadiazines, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, sulphadiazines, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    68-35-9
  • Formula:
    C₁₀H₁₀N₄O₂S
  • Description:
    Sulfadiazine for identification

Specifications

Frequently Bought Together