Meat homogenate - nutrients, NIST-1546A
Catalog # PROONIST-1546A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:4
- Description:Meat homogenate - nutrients
Specifications
About this item
This standard reference material is intended for determining fatty acids, cholestrol, proximates, calories, elements, vitamins and amino acids in canned meat products and similiar materials. The meat homogenate is a mixture of pork and chicken products blended together in a commercial process.
Certifications: Traceable to NIST and SRM numbers.