Impurity and API standard, naphthalen-2-ol (beta-Naphthol)
Catalog # PROOMM0731.01
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:135-19-3
- Description:Naphthalen-2-ol (beta-naphthol)
- Environmentally Preferable:
- Formula:C₁₀H₈O
- Description:Naphthalen-2-ol (beta-naphthol)