Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, quercetin dihydrate, Ph. Eur., RS
Organic reference standards, quercetin dihydrate, Ph. Eur., RS
Catalog # EDQMY0001009
Supplier:  EDQM
CAS Number:  
Organic reference standards, quercetin dihydrate, Ph. Eur., RS
Catalog # EDQMY0001009
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Reference standard
  • CAS No.:
    6151-25-3
  • Description:
    Quercetin dihydrate
  • Seq:
    04
  • Formula:
    C₁₅H₁₄O₉
  • Description:
    Quercetin dihydrate

Specifications

About this item