Order Entry
Austria
ContactUsLinkComponent
Impurity and API standard, 7-chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
Impurity and API standard, 7-chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
Catalog # PROOMM0136.06
CAS Number:  
Impurity and API standard, 7-chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
Catalog # PROOMM0136.06
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    448950-89-8
  • Description:
    7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine
  • Formula:
    C₁₇H₁₃ClN₄
  • Description:
    7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine

Specifications

About this item