Order Entry
Austria
ContactUsLinkComponent
RajHans ChromoSelect medium, modified suitable for microbiology, NutriSelect® Plus, Millipore®
RajHans ChromoSelect medium, modified suitable for microbiology, NutriSelect® Plus, Millipore®
Catalog # SIAL90918-250G
Supplier:  Merck
CAS Number:  
RajHans ChromoSelect medium, modified suitable for microbiology, NutriSelect® Plus, Millipore®
Catalog # SIAL90918-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Description:
    RajHans medium, modified, homogeneous, free flowing powder

Specifications

About this item

RajHans ChromoSelect Medium is recommended for identification and differentiation of Salmonella species from the members of Enterobacteriaceae, especially Proteus species.

  • Final pH: 7,3 ±0,2 (25°C)
  • Appearance: Pinkish yellow to beige coloured