Glucose-6-Phosphate Dehydrogenase
Supplier: Merck
Certificates
About this item
Glucose-6-phosphate dehydrogenase is used to test ketose reductase activity in developing maize endosperm. It catalyses the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway. Also it is a key regulatory enzyme in the first step of the pentose phosphate pathway.
Specifications
- CAS No.:9001-40-5
- Storage conditions:2...8 °C
- Enzyme name:Glucose-6-Phosphate Dehydrogenase
- pH:7,5
Specifications
Frequently Bought Together