Orders LinkContactUsLinkComponent
Prisms
Prisms, equilateral stand, 40×40 mm
Catalog # DIDAPOD010615
Supplier:   DIDALAB
Prisms
Catalog # DIDAPOD010615
Supplier:  DIDALAB
Prisms
Prisms
Prisms, equilateral stand, 40×40 mm
Price
17,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Prism made of standard optical glass with index near to 1,52. Equilateral dispersing prisms are used for wavelength separation applications. Intended for demonstration experiments of light dispersion.
Specifications
  • Description:
    Prisms, equilateral stand, 40×40 mm